Learn More
Fisherbrand™ Isotemp™ Hotplate Products for Science Education
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
$263.00 - $289.00
Specifications
Certifications/Compliance cCSAus
Description Hotplate
Electrical Requirements 100/120 V, 50/60 Hz
Plug Type US style
Frequency 50/60 Hz
View More Specs
Products 2
Catalog Number Size Price Quantity & Availability  
Catalog Number Size Price Quantity & Availability  
S43492 Products for Science Education View Documents 4.25 x 4.25 in. Each for $263.00
Add to cart
S43496 Products for Science Education View Documents 7.25 x 7.25 in. Each for $289.00
Add to cart
Description
Description
Advanced technology
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Specifications
Specifications
cCSAus
100/120 V, 50/60 Hz
50/60 Hz
Ceramic
Hotplate
US style
540°C (Max Temperature)
110/120 V