Learn More
Fisherbrand™ IsoTemp Premium Starter Kit
For limited time, bundle and save up to $900 on select Fisher Scientific™ Isotemp™ Refrigerated/Heated Bath Circulator packages (Models 4100 R20, 6200 R28 and 6200 R35)
 View more versions of this product
Catalog No. 14-259-304
Add to cart
Description
Description
  • Seamless Stainless steel tanks guarantee durability and easy cleaning
  • Robust refrigeration system ensure that samples and application can be cooled quickly
  • Fine-tuned refrigeration metering allows for temperature stability at all times
  • Latest technology, features and options to deliver ideal combination of performance , ease of use and reliability
Specifications
Specifications
60Hz
25.8in.
10.2in.
5.9D x 6.8W x 7.2 in. L
Qty 1 6200 R28 (-28° to 200°C). Qty 1 Sil 180 10L, Qty 1 Plumbing Package and Qty 1 NEScomm software
Force and Suction
80 lb.
19.3in.
19.3in.
±0.05°C
49 x 25.9 x 65.5cm (19.3 x 10.2 x 25.8in.)
750mbar
21L/min.
6200 R28