Learn More
Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer
Fisherbrand™ Isotemp Ultra Low Temperature Freezers combine reliability and performance with cost effective operation and innovative features. Built using an advanced technology platform, refrigeration, microprocessor controls and high quality construction
 View more versions of this product
Catalog No. IUE586FDRK
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Description
Description
Features:
  • Minimal temperature variation and fast door opening recovery
  • Quiet operation
  • Low energy and HVAC costs
  • Envronmentally friendly - utilizes green refrigerants and water blown foam that are SNAP compliant
Warranty and Services
2 years parts and labor + additional 3 yrs. compressor parts.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 Shelves of Racks
2 years parts and labor + additional 3 yrs. compressor parts
Not Available
10 Sliding Drawer Racks (Cat. No. 398329) and 200 Storage Boxes (820109).
500 box
208 V
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For research only