Learn More
Pyramex™ Isotope™ Safety Eyewear
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Mirror
Description
Specifications
Lens Type | H2MAX |
Certifications/Compliance | ANSI Z87.1 ,CSA Z94.3-07 |
Description | Dielectric |
Certifications/Compliance (Europe) | EN-166 |
Lens Tint | Gray |
Frame Color | Black |
Weight (Metric) | 41 g |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |