Learn More
Bio X Cell InVivoMAb Rat IgG2a Control, Anti-Trinitrophenol, 25 MG
Supplier: Bio X Cell BE0089R025MG
The 2A3 monoclonal antibody reacts with trinitrophenol. As trinitrophenol is not expressed by mammals, this antibody is suitable for use as an isotype-matched control for rat IgG2a antibodies in most in vivo and in vitro applications.
- Isotype-matched control for rat IgG2a antibodies
- Suitable for in vivo and in vitro applications
- Unconjugated
- Contains no stabilizers or preservatives
- Purified from cell culture supernatant in an animal-free facility
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.