Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isovaleric anhydride, 95%, Acros Organics

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
1468-39-9
C10H18O3
186.25
FREZLSIGWNCSOQ-UHFFFAOYSA-N
isovaleric anhydride,3-methylbutanoic anhydride,butanoic acid, 3-methyl-, anhydride,butanoic acid, 3-methyl-, 1,1'-anhydride,isovalericanhydride,isovaleriansaureanhydrid,iso-pentanoic anhydride,acmc-209cxm,isovaleric anhydride gc,butanoic acid,3-methyl-,1,1'-anhydride
73847
3-methylbutanoyl 3-methylbutanoate
CC(C)CC(=O)OC(=O)CC(C)C
1468-39-9
C10H18O3
186.25
FREZLSIGWNCSOQ-UHFFFAOYSA-N
isovaleric anhydride,3-methylbutanoic anhydride,butanoic acid, 3-methyl-, anhydride,butanoic acid, 3-methyl-, 1,1'-anhydride,isovalericanhydride,isovaleriansaureanhydrid,iso-pentanoic anhydride,acmc-209cxm,isovaleric anhydride gc,butanoic acid,3-methyl-,1,1'-anhydride
73847
3-methylbutanoyl 3-methylbutanoate
CC(C)CC(=O)OC(=O)CC(C)C
Quantity 100mL
Packaging Glass bottle
Boiling Point 215°C
Density 0.9400g/mL
Flash Point 91°C
Specific Gravity 0.94
Assay Percent Range 94% min
Solubility Information Solubility in water: hydrolyses
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material Isovaleric anhydride
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in original container.

GHS Signal Word: Danger

EINECSNumber : 215-994-3