Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isovalerophenone 99.0+%, TCI America™
Click to view available options
Quantity:
5mL
25mL
Specifications
Boiling Point | 236°C |
Color | Yellow |
Quantity | 25mL |
Formula Weight | 162.23 |
Physical Form | Clear Liquid at 20°C |
Percent Purity | ≥99.0% (GC) |
Chemical Name or Material | Isovalerophenone |
Safety and Handling
TSCA : Yes