Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Isovaleryl-L-carnitine, ≥94.0% (HPLC), MilliporeSigma™ Supelco™

Specifications

Percent Purity ≥94.0% (HPLC)
Grade Analytical Standard
Linear Formula C12H23 NO4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3888
View Documents
MilliporeSigma Supelco
9140310MG
10 mg
Each for $108.90
N/A
11-101-3889
View Documents
MilliporeSigma Supelco
9140350MG
50 mg
Each for $355.50
N/A
Description

Description

Specifications

Specifications

≥94.0% (HPLC)
Analytical Standard
C12H23 NO4
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit