Learn More
Tocris Bioscience™ ISOX DUAL Available on GSA/VA Contract for Federal Government customers only.
Dual BRD4 and CBP/p300 bromodomain inhibitor
$256.00 - $1076.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-061-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5806/10
Each for $256.00
Add to cart
58-065-0 View Documents Tocris Bioscience™
5806/50
Each for $1,076.00
Add to cart
Description
Description
Dual CBP/p300 and BRD4 bromodomain inhibitor (IC50 values are 0.65 and 1.5 μM for CBP and BRD4 respectively). Downregulates IL-6, IL-1β and IFN-β in macrophages.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.