Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoxsuprine Hydrochloride, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
579-56-6
C18H24ClNO3
337.84
QVPSGVSNYPRFAS-UHFFFAOYNA-N
4-{1-hydroxy-2-[(1-phenoxypropan-2-yl)amino]propyl}phenol hydrochloride
Cl.CC(COC1=CC=CC=C1)NC(C)C(O)C1=CC=C(O)C=C1
579-56-6
C18H24ClNO3
337.84
QVPSGVSNYPRFAS-UHFFFAOYNA-N
4-{1-hydroxy-2-[(1-phenoxypropan-2-yl)amino]propyl}phenol hydrochloride
Cl.CC(COC1=CC=CC=C1)NC(C)C(O)C1=CC=C(O)C=C1

I1170, 579-56-6; 34331-89-0, C18H23NO3•HCl

Spectrum™ Isoxsuprine Hydrochloride, USP is a vasodilating agent used for improving blood flow. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.002
Quantity 5 g
CAS Max % 1
Percent Purity 97 to 103%