Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISPD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287639
Description
ISPD Polyclonal specifically detects ISPD in Mouse samples. It is validated for Western Blot.Specifications
ISPD | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
729920 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog, isoprenoid synthase domain containing, isoprenoid synthase domain-containing protein | |
The immunogen is a synthetic peptide directed towards the middle region of mouse ISPD | |
100 μL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction