Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISX 9, Tocris Bioscience™

Catalog Number p-4816934
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
832115-62-5
C11H10N2O2S
234.273
SYENTKHGMVKGAQ-UHFFFAOYSA-N
19582717
N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide
C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3
This item is not returnable. View return policy
832115-62-5
C11H10N2O2S
234.273
SYENTKHGMVKGAQ-UHFFFAOYSA-N
19582717
N-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide
C1CC1NC(=O)C2=NOC(=C2)C3=CC=CS3

Neurogenic agent; induces neuronal differentiation of SVZ progenitors and also induces cardiomyogenic differentiation

ISX 9 is a neurogenic agent. Mediates neuroD reporter gene induction via activation of Ca2+ influx; increases expression of neurogenic differentiation 1 (NeuroD1) transcription factor. Induces neuronal differentiation in human cortical neuronal cells (HCN), adult mouse whole brain (MWB) and subventricular zone (SVZ) progenitors. Also shown to induce cardiomyogenic differentiation of Notch-activated epicardium-derived cells in vitro.

ISX 9 synthesized to Ancillary Material Grade also available.

For more information about how ISX 9 may be used, see our protocol: Transdifferentiating Fibroblasts into Neurons .

Quantity 10 mg
Synonym isx-9, isx 9, n-cyclopropyl-5-thiophen-2-yl isoxazole-3-carboxamide, isoxazole 9 isx-9, neuronal differentiation inducer iii, n-cyclopropyl-5-thiophen-2-yl-1,2-oxazole-3-carboxamide, isoxazole 9, isx9, isoxazole 9 isx9, n-cyclopropyl-5-2-thienyl-3-isoxazolecarboxamide
Formula Weight 234.27
Percent Purity >99%
Chemical Name or Material ISX 9
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit