Learn More
Tocris Bioscience™ IT 901 Available on GSA/VA Contract for Federal Government customers only.
NF-κB c-Rel subunit inhibitor
$205.00 - $848.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-461-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5846/10
Each for $205.00
Add to cart
58-465-0 View Documents Tocris Bioscience™
5846/50
Each for $848.50
Add to cart
Description
Description
c-Rel inhibitor (IC50 = 3 μM). Inhibits IL-2 expression in activated T-cells in vitro. Reduces severity of graft-versus-host disease following hematopoietic stem cell transplant (HSCT) and inhibits growth of human B-cell lymphoma xenografts in mice. Inhibits the oxidative stress response in lymphoma cells.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.