Learn More
ITC Surgicutt™ Bleeding Time Device
Standardized incision depth avoids deep cuts and ensures an incision well above concentrated pain fibers
Manufacturer:  INTERNATIONAL TECHNIDYNE SUN50I
 View more versions of this product
Catalog No. 22-211-509
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Stainless Steel
0.05cm
Avoids deep cuts and ensures an incision well above concentrated pain fibers, used to screen for platelet function
0.098 in.
Newborn
0.019 in.
Yes
Retractable blade
25cm
Bleeding Time Device