Learn More
Selleck Chemical LLCSupplier Diversity Partner Givinostat (ITF2357) 50mg 732302-99-7  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S217050MG
Givinostat (ITF2357) is a potent HDAC inhibitor for maize HD2, HD1B and HD1A with IC50 of 10 nM, 7.5 nM and 16 nM in cell-free assays. Phase 2.
Catalog No. 50-715-5