Learn More
Selleck Chemical LLC
Givinostat (ITF2357) 10mg 732302-99-7
Manufacturer: Selleck Chemical LLC S217010MG
Givinostat (ITF2357) is a potent HDAC inhibitor for maize HD2, HD1B and HD1A with IC50 of 10 nM, 7.5 nM and 16 nM in cell-free assays. Phase 2.