Learn More
Aviva Systems Biology Corp FAM234A Antibody - middle region (ARP50030_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50030_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITFG3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ITFG3. Alternate names: C16orf9, DKFZP761D0211, FLJ32603, gene +108, gs19ApplicationWB. Species reactivity: Human
Catalog No. 50-014-63069