Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2B Mouse anti-Human, Clone: HIP8 (APC), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13901

Catalog No. 89-033-966


Add to cart

Description

Description

Mouse monoclonal antibody raised against human ITGA2B.

  • High specificity
  • High reactivity
Specifications

Specifications

ITGA2B
HIP8
Mouse monoclonal antibody raised against human ITGA2B.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
ITGA2B
Idiopathic thrombocytopenic purpura (IPT) patient platelets.
Protein A/G purification
RUO
Primary
3674
Flow Cytometry, Immunohistochemistry (Frozen)
APC
Flow Cytometry (20 uL/106 cells) Immunohistochemistry (Frozen sections) The optimal working dilution should be determined by the end user.
P08514
CD41|CD41B|GP2B|GPIIb|GTA|HPA3
Mouse
IgG1, kappa
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)