Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA4 Mouse anti-Human, Clone: ALC1/1 (PerCP), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13699

Catalog No. 89-033-764


Add to cart

Description

Description

Mouse monoclonal antibody raised against human ITGA4.

  • High specificity
  • High reactivity
Specifications

Specifications

ITGA4
ALC1/1
Mouse monoclonal antibody raised against human ITGA4.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
ITGA4
Cell preparation of a Jurkat leukemia cell line.
Protein A/G purification
RUO
Primary
3676
Flow Cytometry
PerCP
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
P13612
CD49D|IA4|MGC90518
Mouse
IgG1
100 Reactions
Store in the dark at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)