Learn More
ITGA4 Recombinant Polyclonal Antibody (7HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Supplier: LIFE TECHNOLOGIES 711218
Description
This antibody is predicted to react with Monkey, Horse, Pig Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
ITGA4 Recombinant Polyclonal antibody specifically detects ITGA4 in Human samples. It is validated for Immunocytochemistry.Specifications
ITGA4 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P13612 | |
Rabbit | |
Protein A | |
RUO | |
3676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
7HCLC | |
Unconjugated | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Protein corresponding to human ITGA4 (aa790-1033) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.