Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA5, anti-Human, Clone: M200 (Volociximab), Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00088

Catalog No. 89-032-431


Add to cart

Specifications

Specifications

ITGA5
M200 (Volociximab)
Blocking ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
integrin, alpha 5 (fibronectin receptor, alpha polypeptide)
CD49e|FNRA|VLA5A
Original antibody is raised against heterodimeric alpha 5 beta 1 Fc fusion protein.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
3678|3688
Blocking Assay, ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
ITGA5
IgG, kappa
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit