Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA6 Mouse anti-Human, Clone: 129CT49.6.5, Abnova™

Mouse monoclonal antibody raised against human ITGA6.

Manufacturer:  Abnova Corporation MAB12290

Catalog No. 89-934-387


Add to cart

Description

Description

The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

ITGA6
129CT49.6.5
Mouse monoclonal antibody raised against human ITGA6.
In PBS (0.09% sodium azide)
CD49f/DKFZp686J01244/FLJ18737/ITGA6B/VLA-6
Mouse
IgG1
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:10-1:50) Immunohistochemistry (1:10-1:50) Immunofluorescence (1:10-1:50) Western Blot (1:100-1:16000) The optimal working dilution should be determined by the end user.
Liquid
ITGA6
Recombinant protein corresponding to human ITGA6.
Protein G purification
RUO
Primary
3655
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit