Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGAL Mouse anti-Human, Clone: HI111 (PerCP), Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13793
Description
Mouse monoclonal antibody raised against native human ITGAL.
- High specificity
- High reactivity
Specifications
ITGAL | |
Monoclonal | |
PerCP | |
Blocking Flow Cytometry (20 uL/106 cells) Immunohistochemistry (Frozen sections) Western Blot The optimal working dilution should be determined by the end user. | |
P20701 | |
ITGAL | |
Human CD11a. | |
100 Reactions | |
Primary | |
Human | |
Liquid |
Blocking Assay, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
HI111 | |
Mouse monoclonal antibody raised against native human ITGAL. | |
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
CD11A/LFA-1/LFA1A | |
Mouse | |
Protein A/G purification | |
RUO | |
3683 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction