Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGAL Mouse anti-Human, Mouse, Clone: HI111 (CF-Blue), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13795

Catalog No. 89-033-860


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human ITGAL.

  • High specificity
  • High reactivity
Specifications

Specifications

ITGAL
HI111
Mouse monoclonal antibody raised against native human ITGAL.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
ITGAL
Human CD11a.
Protein A/G purification
RUO
Primary
3683
Blocking Assay, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Unconjugated
Blocking Flow Cytometry (5 uL/106 cells) Immunohistochemistry (Frozen sections) Western Blot The optimal working dilution should be determined by the end user.
P20701
CD11A|LFA-1|LFA1A
Mouse
IgG1, kappa
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)