Learn More
CD11c Mouse anti-Human, PerCP, Clone: BU15, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15803
Catalog No. 50-113-7113
Add to cart
Description
Description
CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
Specifications
Specifications
CD11c
Monoclonal
PerCP
ITGAX
Liquid
ITGAX
Dendritic cells of synovial fluid
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BU15
PBS with 0.2% BSA and 15mM sodium azide
P20702
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Mouse
IgG1
100 tests
4° C
Primary
3687