Learn More
ITGB1, Mouse anti-Human, Clone: 4B7R, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB16399
Catalog No. 89-999-157
Add to cart
Description
Description
Specifications
Specifications
ITGB1
4B7R
Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12)
CD29/FNRB/GPIIA/MDF2/MSK12/VLA-BETA/VLAB
Mouse
IgG1, kappa
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
In 50 mM Na3PO4, 100 mM KCl, 150 mM NaCl, pH 7.5.
Liquid
ITGB1
Human ocular melanoma cell line, V+B2.
50 μg
Store at 4°C. Do not freeze. Open under aseptic conditions.
Primary
3688