Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGB3 Rabbit anti-Human, Unconjugated, Clone: ITGB3/3126R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00564

Catalog No. 89-339-520


Add to cart

Description

Description

ITGB3 Monoclonal antibody specifically detects ITGB3 in Human samples. It is validated for IHC-P.
Specifications

Specifications

ITGB3
ITGB3/3126R
integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61)
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
ITGB3
CD61, GP3A, GPIIIa
Recombinant protein corresponding to amino acids 385-490 of human ITGB3
100 μg
Primary
3690
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit