Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRB Mouse anti-Human, APC, Clone: 18A2, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15719

Catalog No. 50-113-7075


Add to cart

Description

Description

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, alpha and beta coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, alpha while PDGF BB can bind and activate to both PDGFR, alpha and beta.
Specifications

Specifications

PDGFRB
Monoclonal
APC
PDGFRB
Liquid
PDGFRB
CD140B-transfected NIH-3T3 cells
Size-exclusion chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
18A2
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P09619
PDGF Receptor beta, JTK12, PDGFR, PDGFR1, CD140 antigen-like family member B, PDGF-R-beta, beta-type platelet-derived growth factor receptor
Mouse
IgG1
100 tests
4° C, store in dark, DO NOT FREEZE!
Primary
5159
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit