Learn More
Abnova™ ITIH1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003697T01
Catalog No. 89-949-343
Add to cart
Description
Description
Specifications
Specifications
293T
NM_002215.1
pCMV-ITIH1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
ITIH1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 100.32kDa
Transient overexpression cell lysate was tested with anti-ITIH1 antibody (Mfr. No. H00003697-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human