Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ITK, GST Tag Recombinant Protein
ITK Recombinant Human Protein
Supplier: LIFE TECHNOLOGIES PV3875
Description
ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation.Specifications
NP_005537 | |
114294, 3702 | |
10μg | |
TEC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
IL2-inducible T-cell kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
99.9 kDa | |
ITK, EMT, LYK, PSCTK2 | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
ITK | |
Baculovirus | |
Full Length, Recombinant, Cytoplasmic | |
Cytoplasmic Tyrosine Kinases |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction