Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITK Recombinant Human Protein Non-distribution product as customer accommodation.

ITK Recombinant Human Protein

$431.00

Specifications

Accession Number NP_005537
Expression System Baculovirus
Gene Alias ITK, EMT, LYK, PSCTK2
KinaseFamily TEC Kinase Family
Molecular Weight 99.9 kDa
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3875 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
PV3875
10μg Each for $431.00
Add to cart
 
Description

Description

ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation.
Specifications

Specifications

NP_005537
ITK, EMT, LYK, PSCTK2
99.9 kDa
GST Tag
Baculovirus
TEC Kinase Family
Kinase Assay

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit