Learn More
ITL Corporation SampLok™ Tube Holders
Multi-sample tube holder to collect blood samples into vacuum test tubes and long neck blood culture bottles
Manufacturer: ITL Corporation A100500
Description
- Ready-to-use
- Latex free
- Translucent for full visibility
- Full length holder reduces potential exposure to sampling needle
A100500
- Pre-attached luer
- Sterile, individual package
- Integrated safety lid to reduce risk of potential needlestick injury
Specifications
Sterile | |
Tube Holder | |
Translucent |
For collect Blood samples into vacuum test tubes and long neck Blood culture bottles | |
Yes | |
Preattached luer |