Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL Corporation SampLok™ Tube Holders

Multi-sample tube holder to collect blood samples into vacuum test tubes and long neck blood culture bottles

$203.00 - $324.00

Specifications

Color Translucent
Product Type Tube Holder
For Use With (Application) For collect Blood samples into vacuum test tubes and long neck Blood culture bottles
Latex Free Yes
View More Specs

Products 2
Catalog Number Mfr. No. Sterility Includes Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Includes Price Quantity & Availability  
21-010-220
View Documents Promotion Details
ITL Corporation
A100422
Non-sterile Integrated safety lid
Case of 700 for $203.00
Add to cart
 
21-010-221
View Documents Promotion Details
ITL Corporation
A100500
Sterile Preattached luer
Case of 425 for $324.00
Add to cart
 
Description

Description

  • Ready-to-use
  • Latex free
  • Translucent for full visibility
  • Full length holder reduces potential exposure to sampling needle

A100500

  • Pre-attached luer
  • Sterile, individual package
A100422
  • Integrated safety lid to reduce risk of potential needlestick injury

Specifications

Specifications

Translucent
Tube Holder
For collect Blood samples into vacuum test tubes and long neck Blood culture bottles
Yes
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit