Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITM2A Monoclonal Antibody (OTI10C11), OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811876


Description
ITM2A Monoclonal antibody specifically detects ITM2A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ITM2A | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P40938, Q8R323 | |
Mouse | |
Protein A/G | |
RUO | |
288414, 5983, 69263 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI10C11 | |
Unconjugated | |
RFC3 | |
AW108051; BRICD2A; BRICHOS domain containing 2A; E25; E25A; Integral membrane protein 2A; Itm2; ITM2A; Protein E25; UNQ603/PRO1189 | |
Full length human recombinant protein of human RFC3 (NP_002906) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction