Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITM2B Mouse anti-Human, PE-Cyanine7, Clone: OTI1C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271460PECY7

Catalog No. N271460PEC7


Add to cart

Description

Description

ITM2B Monoclonal antibody specifically detects ITM2B in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ITM2B
OTI1C11
Flow Cytometry
Affinity Purified, Conjugated
ITM2B
Full length recombinant protein of human ITM2B (NP_068839) produced in HEK293T cells.
Immunogen affinity purified
Neuroscience, Signal Transduction
Primary
9445.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
ABRI, ABri/ADan amyloid peptide, BRI2, BRICD2B, BRICHOS domain containing 2B, BRIFBD, E25B, E3-16, integral membrane protein 2B, Protein E25B, Transmembrane protein BRI
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit