Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITSN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545703
Description
Peptide sequence: TFTEGEEILV TQKDGEWWTG SIGDRSGIFP SNYVKPKDQE SFGSASKSGA Sequence homology: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 93%; Rat: 79%.
This gene encodes a cytoplasmic protein which contains SH3 domains. This protein is a member of a family of proteins involved in clathrin-mediated endocytosis. Intersectin 2 is thought to regulate the formation of clathrin-coated vesicles and also may function in the induction of T cell antigen receptor (TCR) endocytosis. Alternatively spliced transcript variants have been found for this gene that encode three distinct isoforms. Additional variants have been found but their full length nature has not been determined.
Specifications
ITSN2 | |
Polyclonal | |
Unconjugated | |
ITSN2 | |
AI327390; EH and SH3 domains protein 2; EH domain and SH3 domain regulator of endocytosis 2; Eh domain, SH3 domain regulator of endocytosis 2; Ese2; intersectin 2; intersectin-2; Intersectin-2-like protein; ITSN2; KIAA1256; LOW QUALITY PROTEIN: intersectin-2; mKIAA1256; PRO2015; SH3 domain protein 1B; SH3 domain-containing protein 1B; SH3D1B; Sh3p18; SH3P18-like WASP-associated protein; SWA; SWAP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
50618 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZM3 | |
ITSN2 | |
Synthetic peptide directed towards the C-terminal of human ITSN2 (aa 1001-1050). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction