Texwipe™ Mini AlphaMop™ Replacement Microdenier Mop Covers
Manufacturer: Texwipe™ TX7114M
 View more versions of this product
Catalog No. 19-139-875
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Maximum removal of small and micro contaminants and used in cleanrooms, disinfecting with quat-based cleaners, pharmaceuticals, biologics, medical device
    Material 100% Polyester Fiber Microdenier
    Product Type Microdenier Mop Covers
    Sterile No
  • Description & Specifications
Related Products
Product Content Feedback