Ivachtin, Tocris

Potent caspase-3 inhibitor

Manufacturer: R&D Systems 2636/1

Quantity 1mg

 View more versions of this product

Catalog No. 26-361

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C20H21N3O7S
    Formula Weight 447.46
    Synonym 2-[4-Methyl-8-(morpholin-4-ylsulfonyl)-1,3-dioxo-1,3-dihydro-2H-pyrrolo[3,4-c]quinolin-2-yl]ethyl acetate
    Chemical Name or Material Ivachtin

    Specifications

    Formula Weight 447.46
    Quantity 1mg
    Synonym 2-[4-Methyl-8-(morpholin-4-ylsulfonyl)-1,3-dioxo-1,3-dihydro-2H-pyrrolo[3,4-c]quinolin-2-yl]ethyl acetate
    Assay Percent Range >99%
    Molecular Formula C20H21N3O7S
    CAS 745046-84-8
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material Ivachtin

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit