Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ivermectin, For Vet Use, USP, 95-102%, Spectrum™

I3012, 70288-86-7

$118.00 - $17221.00

Specifications

CAS 70288-86-7
CAS Max % 1
Percent Purity 95 to 102%
UN Number UN2811
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-603-596 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I30121GM
1 g Amber Glass Bottle Each for $118.00
Add to cart
 
18-603-597 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I30125GM
5 g Amber Glass Bottle Each for $432.00
Add to cart
 
18-603-598 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I301225GM
25 g Amber Glass Bottle Each for $1,212.00
Add to cart
 
18-603-599 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I3012100GM
100 g Amber Glass Bottle Each for $2,777.00
Add to cart
 
18-603-600 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I3012500GM
500 g Amber Glass Bottle Each for $4,280.00
Add to cart
 
18-603-601 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I30121KGBL
1 kg Amber Glass Bottle Each for $5,386.00
Add to cart
 
18-603-602 View Documents Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
I301220KGBL
20 kg Fiber Drum Each for $17,221.00
Add to cart
 
Description

Description

Spectrum™ Ivermectin, For Vet Use, USP
Specifications

Specifications

70288-86-7
1
95 to 102%
UN2811
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit