Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IZUMO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572869
Description
IZUMO1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The sperm-specific protein Izumo, named for a Japanese shrine dedicated to marriage, is essential for sperm-egg plasma membrane binding and fusion.The sperm-specific protein Izumo, named for a Japanese shrine dedicated to marriage, is essential for sperm-egg plasma membrane binding and fusion.
Specifications
IZUMO1 | |
Polyclonal | |
Unconjugated | |
Izumo1 | |
1700058F15Rik; Izumo; izumo sperm-egg fusion 1; izumo sperm-egg fusion protein 1; IZUMO1; MGC34799; OBF; oocyte binding/fusion factor; Sperm-specific protein izumo | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284359 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8IYV9 | |
Izumo1 | |
IZUMO1 antibody for an 18 amino acid peptide near the center of human IZUMO1. The immunogen is within amino acids 170 - 220 of IZUMO1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction