Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
J Chain Mouse anti-Human, Clone: Mc19-9, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | J Chain |
---|---|
Clone | Mc19-9 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 20 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 20 ug/ml, Immunohistochemistry-Frozen 1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10064825
|
Novus Biologicals
NB10064825 |
0.1 mg |
Each for $436.00
|
N/A |
NB017652
|
Novus Biologicals
NB100648250.025MG |
0.025 mg |
Each for $176.85
|
N/A |
Description
J Chain Monoclonal specifically detects J Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
J Chain | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human | |
IgG1 | |
Protein A purified | |
NB100-64825 reacts with human J chain as tested by ELISA using purified human Ig fragments as the solid phase target antigen. The antibody does not detect J chain in intact IgM in ELISA assays due to steric hindrance. In Western blotting techniques NB100-64825 detects J chain in IgM only in reducing conditions. |
Mc19-9 | |
Western Blot 1:100-1:2000, ELISA 20 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 20 ug/ml, Immunohistochemistry-Frozen 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human J Chain | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title