JAC Medical Accessories for Mobile Phlebotomy Carts

Manufacturer: JAC Medical ProductsSupplier Diversity Partner JAC2007

 View more versions of this product

Catalog No. 22-038-4563

  Check Availability
Add to Cart

    Specifications

    Color White
    For Use With (Application) Extra handling control of cart from the side
    For Use With (Equipment) Phlebotomy Carts
    Product Type Side handle bar
    Includes Sponge grip
    Material White powder coated metal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit