JAC Medical JAC-Advance Large Phlebotomy Tray Units

For large to medium volume blood draws

Manufacturer: JAC Medical ProductsSupplier Diversity Partner JAC 102

 View more versions of this product

Catalog No. 22-038441

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    Color Blue
    For Use With (Application) For large to medium volume Blood draws
    For Use With (Equipment) For drawing 40+ tubes
    Height (English) Handle 6 in.
    Height (Metric) Handle 15.24cm
    Includes Packet holder, gauze box, accessory box, supply box, glove holder, mini utility box, isolation box with two needle holders, hold 32VAC tubes each, centered handle
    Size Large
    Material Polypropylene
    Product Type Phlebotomy Tray Unit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit