Kimberly Clark™ Professional Jackson Safety™ G29 Solvent Gloves

Provide the dexterity of thin mil gloves with greater durability for superior protection. Kimberly Clark™ Professional Jackson Safety™ G29 Solvent Gloves feature neoprene blend construction to protect against solvents and won't break apart after 15 minutes of work like thin mil gloves.

Manufacturer: Kimberly Clark™ Professional 49827

 View more versions of this product

Catalog No. 19-167-725

  Check Availability
Add to Cart

    Specifications

    Thickness 12 mil (finger)
    9 mil (palm)
    Finger Style Textured
    Glove Cuff Style Beaded
    Hand Ambidextrous
    For Use With (Application) Protection from solvents
    Length (English) 12 in.
    Latex Free Yes
    Material Neoprene blend
    Powdered No
    Size 11 (2XL)
    Color Blue

    • Neoprene blend construction designed for protection from solbents
    • Textured fingertips for wet or dry grip
    • Not made with natural rubber latex
    • Silicone-free
    • Powder-free
    • Ambidextrous for greater efficiency
    • Highest level of dexterity per EN420:2003 standard
    • Available in sizes 6(XS) - 11 (2XL)
    Common Applications:
    • Handling hydraulic fluids
    • Working with solvents such as Frekote, IPA, and Varson
    • Wiping down parts and machinery with solvents
    • Janitorial cleaning

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit