Learn More
Kimberly-Clark Professional™ Jackson Safety™ G29 Solvent Gloves
Provide the dexterity of thin mil gloves with greater durability for superior protection. Kimberly Clark™ Professional Jackson Safety™ G29 Solvent Gloves feature neoprene blend construction to protect against solvents and won't break apart after 15 minutes of work like thin mil gloves.
Manufacturer:  Kimberly-Clark Professional™ 49826
 View more versions of this product
Catalog No. 19-167-724
Add to cart
Description
Description
  • Neoprene blend construction designed for protection from solbents
  • Textured fingertips for wet or dry grip
  • Not made with natural rubber latex
  • Silicone-free
  • Powder-free
  • Ambidextrous for greater efficiency
  • Highest level of dexterity per EN420:2003 standard
  • Available in sizes 6(XS) - 11 (2XL)
Common Applications:
  • Handling hydraulic fluids
  • Working with solvents such as Frekote, IPA, and Varson
  • Wiping down parts and machinery with solvents
  • Janitorial cleaning
Compliance
Meets EN 388, EN 374-2 and EN 420-2003 standards.
Specifications
Specifications
Textured
Ambidextrous
Yes
10, X-Large
Powder-free
12 in.
Beaded/Rolled
Protection from solvents
Neoprene Blend
12 mil (finger)
9 mil (palm)
Blue