Learn More
Kimberly-Clark™ Professional Jackson Safety™ G80 Chemical Resistant Neoprene Gloves Available on GSA/VA Contract for Federal Government customers only.
Yellow and blue, hand-specific gloves providing reliable protection against chemical and basic mechanical hazards.
$69.30 - $69.50
Specifications
Color Yellow and Blue
Dimensions (L x W) 42.4 x 16.8cm
Length (Metric) 42.4cm
Material Neoprene
Product Type Gloves
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-006-228 View Documents Kimberly-Clark™ Professional
38741
7 Pack of 12 for $69.50
Case of 5 PK for $294.94
Add to cart
19-006-229 View Documents Kimberly-Clark™ Professional
38742
8 Pack of 12 for $69.50
Case of 5 PK for $294.94
Add to cart
19-006-230 View Documents Kimberly-Clark™ Professional
38743
9 Pack of 12 for $69.50
Case of 5 PK for $294.94
Add to cart
19-006-231 Available on GSA/VA Contract for Federal Government customers only. View Documents Kimberly-Clark™ Professional
38744
10 Pack of 12 for $69.30
Case of 5 PK for $296.00
Add to cart
Description
Description
  • Suitable for various applications and offer reliable protection against wide range of chemicals
  • Ideal for: providing PPE Category 3 protection, basic mechanical protection in many different segments when handling chemicals, oils, grease, acids, caustics or solvents
  • Hand specific gloves of 0.70mm thickness
  • Made of neoprene over latex formulation to maximize protection
  • Enhances flexibility with straight cuff and honeycomb structure on the palm to provide good grip in wet and dry conditions
  • Cotton flocked liner for easy donning and extra comfort in extended use
  • Chlorinated for low extractable proteins and residual chemicals
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yellow and Blue
42.4cm
Gloves
0.70mm
42.4 x 16.8cm
Neoprene
5.04kg