Learn More
Kimberly-Clark Professional™ Jackson Safety™ V20 Purity™ Safety Glasses Available on GSA/VA Contract for Federal Government customers only.
Economical yet stylish eye protection.
Manufacturer:  Kimberly-Clark Professional™ 25649
 View more versions of this product
Catalog No. 19-170-102
Add to cart
Description
Description
  • Comfortable, padded temples for enhanced comfort
  • Clear lenses allow maximum visible light transmission; ideal for indoor use
  • Anti-Fog coating to prevent fogging; ideal for humid environments and temperature variances
  • Hardcoated lenses for scratch resistance and longer life
  • Lenses are made of a polycarbonate material that provides 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1+ standards for high impact (25654-00 meets ANSI Z87.1+2010 and 25654-01 meets ANSI Z87.1+2015) and certified to meet the requirements of CSA Z94.3
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ANSI Z87.1: 2010; EN 166: 2001; CSA Z94.3:2007; AS/NZS 1337.1:2010
Polycarbonate
Black
Amber
Polycarbonate
Amber
Ratcheted