Learn More
Sklar InstrumentsSupplier Diversity Partner TC Jacobson Needle Holder with Diamond Coated Jaw, Sklar, 8-1/2 in., Straight, Serrated, with Lock. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218134
TC Jacobson Needle Holder with Diamond Coated Jaw, Sklar, 8-1/2 in., Straight, Serrated, with Lock. Sklar's Jacobson Tungsten Carbide Microvascular Needle Holder is primarily used for suturing in microvascular and cardiovascular procedures. It may also be known as a Jacobson-Castroviejo Needle Holder. The finely cross-serrated, tapered jaws are able to securely hold very fine sutures. Serrations on TC instruments are designed to stay sharp for a significantly longer period of time than standard instruments. Sklar also permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with micro-profile, cross-serrated jaws and an overall length of 8-1/2 inches. It also features a locking mechanism.
Catalog No. 50-143-7799
Due to product restrictions, please to purchase or view availability for this product.