Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jagged 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | Jagged 2 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15828420
|
Novus Biologicals
NBP15828420UL |
20 μL |
Each for $152.22
|
|
NBP158284
|
Novus Biologicals
NBP158284 |
100 μL |
Each for $436.00
|
|
Description
Jagged 2 Polyclonal specifically detects Jagged 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Jagged 2 | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
HJ2, jagged 2, Jagged2, protein jagged-2, SER2 | |
JAG2 | |
IgG | |
Affinity Purified |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Stem Cell Signaling Pathway | |
Q9Y219 | |
3714 | |
Synthetic peptides corresponding to JAG2(jagged 2) The peptide sequence was selected from the N terminal of JAG2. Peptide sequence RAQGRGRLPRRLLLLLALWVQAARPMGYFELQLSALRNVNGELLSGACCD. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title