Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jak2 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI7D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271056AF405

Catalog No. N271056A405


Add to cart

Description

Description

Jak2 Monoclonal antibody specifically detects Jak2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Jak2
OTI7D12
Immunohistochemistry
Affinity Purified, Conjugated
JAK2
Recombinant protein fragment corresponding to amino acids 297-596 of human Jak2 (NP_004963) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary
3717.0
Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit